Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzophenone-2 | 131-55-5 | MFCD00002278 | 99.7% | 246.22 g·mol⁻¹ | C13H10O5 | 100 G
SDP

Catalog No. 5000300177
Encompass_Preferred

Benzophenone-2 is a tetrahydroxybenzophenone used as an organic UV filter and a research probe to study nuclear receptor activity, including pregnane X receptor (PXR) and estrogen receptor α (ERα). Supplied as a high-purity solid with documented storage, handling, and quality documentation for laboratory research use.

  • High-purity solid (99.66%).
  • Used as a UV filter and probe for pregnane X receptor (PXR) and estrogen receptor α (ERα) studies.
  • Molecular formula C13H10O5; molecular weight 246.22 g·mol⁻¹.
  • Appearance light yellow solid; supplied in a 100 g pack.
  • Storage: room temperature under nitrogen; in solution: -80°C (2 years), -20°C (1 year).
  • Documentation available: datasheet, SDS, and COA.

Catalog No. 50-003-00177 Supplier Medchemexpress LLC Supplier No. HYW036120100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzophenone-2 is a tetrahydroxybenzophenone used as an organic UV filter and a research probe to study nuclear receptor activity, including pregnane X receptor (PXR) and estrogen receptor α (ERα). Supplied as a high-purity solid with documented storage, handling, and quality documentation for laboratory research use.

  • High-purity solid (99.66%).
  • Used as a UV filter and probe for pregnane X receptor (PXR) and estrogen receptor α (ERα) studies.
  • Molecular formula C13H10O5; molecular weight 246.22 g·mol⁻¹.
  • Appearance light yellow solid; supplied in a 100 g pack.
  • Storage: room temperature under nitrogen; in solution: -80°C (2 years), -20°C (1 year).
  • Documentation available: datasheet, SDS, and COA.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.