Learn More
Medchemexpress LLC Benzophenone-2 (2,2′,4,4′-tetrahydroxybenzophenone) | 131-55-5 | 99.7% | 246.22 g/mol | C13H10O5 | 1 KG
SDP
Catalog No. 5000299561
Encompass_Preferred
Benzophenone-2 (2,2′,4,4′-tetrahydroxybenzophenone; CAS 131-55-5) is a benzophenone-class UV filter used in laboratory research on UV-absorbing compounds and xenobiotic metabolism. It is provided as a reagent for research use only and has reported activity modulating cytochrome P450 expression via pregnane X receptor activation.
  • High purity: 99.66%
  • Molecular formula: C13H10O5; molecular weight: 246.22 g/mol
  • Suitable for UV filter and metabolism research
  • Available in multiple package sizes including 1 kg
  • Provided for research use only
Catalog No. 50-002-99561 Supplier Medchemexpress LLC Supplier No. HYW0361201KG
Benzophenone-2 (2,2′,4,4′-tetrahydroxybenzophenone; CAS 131-55-5) is a benzophenone-class UV filter used in laboratory research on UV-absorbing compounds and xenobiotic metabolism. It is provided as a reagent for research use only and has reported activity modulating cytochrome P450 expression via pregnane X receptor activation.
  • High purity: 99.66%
  • Molecular formula: C13H10O5; molecular weight: 246.22 g/mol
  • Suitable for UV filter and metabolism research
  • Available in multiple package sizes including 1 kg
  • Provided for research use only