Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Benzophenone
SCI_ED

Catalog No. S25664B Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Solid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25664B 100 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25664B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 119-61-9
Molecular Formula C13H10O
Molecular Weight (g/mol) 182.222
InChI Key RWCCWEUUXYIKHB-UHFFFAOYSA-N
Synonym benzophenone, diphenyl ketone, benzoylbenzene, phenyl ketone, methanone, diphenyl, alpha-oxoditane, ketone, diphenyl, benzene, benzoyl, alpha-oxodiphenylmethane, diphenylketone
PubChem CID 3102
ChEBI CHEBI:41308
IUPAC Name diphenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=C2

Specifications

Color White
Density 1.11g/mL
Assay Percent Range >95%
Packaging Solid
Linear Formula (C6H5)2CO
Quantity 100 g
Formula Weight 182.22
Chemical Name or Material Benzophenone
Safety handling Check Alert storage code Red
ShelfLife 2 years
DOTInformation DOT Class 9, III, : Miscellaneous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.