Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzophenone

Catalog No. B93002KGA Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
500 g
1 kg
2 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B93002KGA 500 g
B930025GA 1 kg
B93001KGA 2 kg
B9300500GA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. B93002KGA Supplier Sigma Aldrich Supplier No. B93002KGA
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Benzophenone

Specifications

CAS 119-61-9
Melting Point 47°C to 51°C
Boiling Point 305°C
Molecular Formula C13H10O
Linear Formula (C6H5)2CO
Quantity 500 g
MDL Number MFCD00003076
Synonym Diphenyl ketone; Diphenylmethanone; NSC 8077
Molecular Weight (g/mol) 182.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.