Learn More
Medchemexpress LLC Benzophenone-d10 | 22583-75-1 | MFCD00143675 | 99.6% | 192.28 g·mol⁻1 | C13D10O | 100mg
SDP
Catalog No. 5000268945
Encompass_Preferred
Benzophenone-d10 is the deuterium labeled Benzophenone (HY-Y0546) Benzophenone is an endogenous metabolite Benzophenone is a photosensitizer that absorbs UV light transfers its energy to DNA and triggers DNA damage Benzophenone can be used as the fragrance enhancer ultraviolet curing agent additive in plastics or the flavor ingredient Benzophenone can also be used in the manufacturing of insecticides agricultural chemicals hypnotics antihistamines and other pharmaceuticals Benzophenone exhibits certain carcinogenicity in mouse/rat models under long term exposure Benzophenone exhibits estrogenic activity[1][2]
Catalog No. 50-002-68945 Supplier Medchemexpress LLC Supplier No. HYY0546S100MG
Benzophenone-d10 is the deuterium labeled Benzophenone (HY-Y0546) Benzophenone is an endogenous metabolite Benzophenone is a photosensitizer that absorbs UV light transfers its energy to DNA and triggers DNA damage Benzophenone can be used as the fragrance enhancer ultraviolet curing agent additive in plastics or the flavor ingredient Benzophenone can also be used in the manufacturing of insecticides agricultural chemicals hypnotics antihistamines and other pharmaceuticals Benzophenone exhibits certain carcinogenicity in mouse/rat models under long term exposure Benzophenone exhibits estrogenic activity[1][2]