Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzophenone imine, 97%, stabilized

Catalog No. AC368590250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC368590250 25 g Glass bottle
AC368590050 5 g Glass bottle
AC368591000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC368590250 Supplier Thermo Scientific Chemicals Supplier No. 368590250
Only null left
Add to Cart

CAS: 1013-88-3 | C13H11N | 181.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1013-88-3 , 123-31-9
Molecular Formula C13H11N
Molecular Weight (g/mol) 181.23
MDL Number MFCD00001760
InChI Key SXZIXHOMFPUIRK-UHFFFAOYSA-N
Synonym benzophenone imine, benzophenoneimine, benzhydrylideneamine, 1,1-diphenylmethanimine, unii-ejj21na7vi, benzhydrylimine, benzophenonimine, diphenylmethanimineimine, 1,1-diphenylmethylimine, benzenemethanimine, .alpha.-phenyl
PubChem CID 136809
IUPAC Name diphenylmethanimine
SMILES C1=CC=C(C=C1)C(=N)C2=CC=CC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -30°C
Color Brown-Yellow to Colorless
Density 1.0800g/mL
Boiling Point 216°C (75.0 mmHg)
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.6170 to 1.619
Linear Formula (C6H5)2C=NH
Quantity 25 g
Beilstein 07, IV, 1365
Specific Gravity 1.08
Merck Index 15, 1102
Solubility Information Solubility in water: 0.3g/L (25°C). Other solubilities: better soluble in aq. solution at lower ph values,,very soluble in diethyl ether
Stabilizer approx. 0.02% w/w hydroquinone
Viscosity 33,4 mPa.s (20°C)
Formula Weight 181.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Benzophenone imine, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.