Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzophenone Imine, Sigma-Aldrich

Catalog No. 01259018 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-018 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-259-018 Supplier MilliporeSigma Supelco Supplier No. 29373325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzophenone imine is useful for the preparation of nitrile yield dimers.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1013-88-3
Molecular Weight (g/mol) 181.23 g/mol

Specifications

Boiling Point 151°C to 153°C
Quantity 25 g
Percent Purity 95%
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.