Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzophenone, MilliporeSigma™ Supelco™

Catalog No. 111020251 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0251 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-102-0251 Supplier MilliporeSigma Supelco Supplier No. 442842
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 119-61-9
Vapor Pressure 1 mmHg (108°C)
Flash Point 138°C
Physical Form Neat
Linear Formula (C6H5)2 CO
MDL Number MFCD00003076
Melting Point 47°C to 51°C
Boiling Point 305°C
Synonym Diphenyl ketone; Diphenylmethanone; NSC 8077
Type Food and Cosmetic Component Standard
Quantity 500 mg
Formula Weight 182.22
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.