Learn More
Medchemexpress LLC HY-100511 10mg Medchemexpress, Benzophenonetetracarboxylic acid CAS:2479-49-4 Purity:>98%
SDP
Catalog No. 501931861
Encompass_Preferred
Medchemexpress, HY-100511 10mg Benzophenonetetracarboxylic acid CAS:2479-49-4 Benzophenonetetracarboxylic acid (3,3',4,4'-Benzophenonetetracarboxylic acid) is particularly useful in the preparation of high performance polyimides and also useful as curing agents for epoxy resins [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1861 Supplier Medchemexpress LLC Supplier No. HY10051110MG
Medchemexpress, HY-100511 10mg Benzophenonetetracarboxylic acid CAS:2479-49-4 Benzophenonetetracarboxylic acid (3,3',4,4'-Benzophenonetetracarboxylic acid) is particularly useful in the preparation of high performance polyimides and also useful as curing agents for epoxy resins [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.