Learn More
Medchemexpress LLC Benzophenonetetracarboxylic dianhydride | 2421-28-5 | 99.5% | 322.23 | 100 G
Supplier: Medchemexpress LLC HYW009169100G
Benzophenonetetracarboxylic dianhydride (BTDA) is extensively utilized in the creation of high-performance polyimides and other polymeric materials. It reacts with various diamine compounds to produce oligoimide chains, which can then modify graphene oxide (Go) and enhance its properties. It is intended for research use only.
- Forms oligoimide chains to modify graphene oxide
- Enhances properties of graphene oxide
- Suitable for synthesis of high-performance polyimides
- Used in other polymer materials