Learn More
Medchemexpress LLC Benzophenonetetracarboxylic dianhydride | 2421-28-5 | 99.5% | 322.23 | 500 G
SDP
Catalog No. 5000366865
Encompass_Preferred
Benzophenonetetracarboxylic dianhydride is widely used in the synthesis of high-performance polyimides and other polymer materials. It reacts with various diamine compounds to form oligoimide chains, which can modify graphene oxide (Go) and enhance its properties.
  • The product is for research use only.
  • Appears as a white to light yellow solid.
  • Store at 4°C, sealed and away from moisture.
  • When in solvent, store at -80°C for 6 months; -20°C for 1 month in sealed storage away from moisture.
  • ¹H NMR spectrum is consistent with its structure.
  • Tested and complies with given specifications.
Catalog No. 50-003-66865 Supplier Medchemexpress LLC Supplier No. HYW009169500G
Benzophenonetetracarboxylic dianhydride is widely used in the synthesis of high-performance polyimides and other polymer materials. It reacts with various diamine compounds to form oligoimide chains, which can modify graphene oxide (Go) and enhance its properties.
  • The product is for research use only.
  • Appears as a white to light yellow solid.
  • Store at 4°C, sealed and away from moisture.
  • When in solvent, store at -80°C for 6 months; -20°C for 1 month in sealed storage away from moisture.
  • ¹H NMR spectrum is consistent with its structure.
  • Tested and complies with given specifications.