Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzopinacol, 98%

Catalog No. AAA1142122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1142122 100 g
AAA1142114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1142122 Supplier Thermo Scientific Chemicals Supplier No. A1142122
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 464-72-2 | C26H22O2 | 366.46 g/mol

Benzopinacol is a catalyst of the formation of unsaturated polyesters. It is also used as an initiator of polymerisation by free radicals. It is also used as an organic synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications: Benzopinacol is a catalyst of the formation of unsaturated polyesters. It is also used as an initiator of polymerisation by free radicals. It is also used as an organic synthesis intermediate.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 464-72-2
Molecular Formula C26H22O2
Molecular Weight (g/mol) 366.46
MDL Number MFCD00004448
InChI Key MFEWNFVBWPABCX-UHFFFAOYSA-N
Synonym benzopinacol, benzopinacole, benzopinacone, benzpinacol, benzpinacone, tetraphenylethylene glycol, tetraphenyl-1,2-ethanediol, benzophenone pinacol, 1,1,2,2-tetraphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2,2-tetraphenyl
PubChem CID 94766
IUPAC Name 1,1,2,2-tetraphenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(C3=CC=CC=C3)(C4=CC=CC=C4)O)O

Specifications

Melting Point 174°C to 180°C
Assay Percent Range 98%
Quantity 100 g
Beilstein 2061947
Solubility Information It is soluble in ethanol,ether,acetone insoluble in carbon disulfide and water.
Formula Weight 366.46
Chemical Name or Material Benzopinacol
EINECSNumber 207-356-8
RTECSNumber KI2750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.