Learn More
TARGETMOL CHEMICALS INC BENZOQUINAMIDE HYDROCHLO 5MG
SDP
Catalog No. 502578942
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Benzoquinamide hydrochloride (3-carbamoyl-NN-diethyl-1346711b-hexahydro-911-dimethoxy-2H-benzo[a]quinolizin-2-yl acetate hydrochloride) is an antiemetic agent and increases intra-arterial pressure due to the increased peripheral vascular resistance. purity: 99%
Catalog No. 50-257-8942 Supplier TARGETMOL CHEMICALS INC Supplier No. T98815MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Benzoquinamide hydrochloride (3-carbamoyl-NN-diethyl-1346711b-hexahydro-911-dimethoxy-2H-benzo[a]quinolizin-2-yl acetate hydrochloride) is an antiemetic agent and increases intra-arterial pressure due to the increased peripheral vascular resistance. purity: 99%