Learn More
U.S. Pharmacopeia Benzothiadiazine Related Compound A, 121-30-2, MFCD00007933, 100 mg
Supplier:  U.S. Pharmacopeia 1057507
Encompass
Linear Formula: H2NC6H2(Cl)(SO2NH2)2, Molecular Weight: 285.73, mp: 257-261 °C, Synonym: 3-Chloroaniline-4,6-disulfonamide, 4-Amino-6-chlorobenzene-1,3-disulfonamide.
Catalog No. NC1160868