Learn More
Medchemexpress LLC Benzothiazole-5-boronic acid pinacol ester | 1073354-91-2 | MFCD09260439 | ≥98.0% | 261.15 g·mol⁻1 | C13H16BNO2S | 250 MG
SDP
Catalog No. 5000302591
Encompass_Preferred
Benzothiazole-5-boronic acid pinacol ester is a boronic ester reagent used as the boronate coupling partner in Suzuki-Miyaura cross-coupling reactions. It serves as a versatile building block for constructing biaryl and heteroaryl motifs in medicinal chemistry and organic synthesis and is supplied for research applications with analytical characterization.
Catalog No. 50-003-02591 Supplier Medchemexpress LLC Supplier No. 250MGHYW019633
Benzothiazole-5-boronic acid pinacol ester is a boronic ester reagent used as the boronate coupling partner in Suzuki-Miyaura cross-coupling reactions. It serves as a versatile building block for constructing biaryl and heteroaryl motifs in medicinal chemistry and organic synthesis and is supplied for research applications with analytical characterization.