Learn More
Medchemexpress LLC Benzothiazole-5-boronic acid pinacol ester | 1073354-91-2 | MFCD09260439 | ≥98.0% | 261.15 g/mol | C13H16BNO2S | 100 G
SDP
Catalog No. 5000300947
Encompass_Preferred
Benzothiazole-5-boronic acid pinacol ester is an organoboron reagent used as the boronate partner in Suzuki-Miyaura cross-coupling reactions. It is commonly used in medicinal chemistry and synthetic organic research and should be handled and stored according to manufacturer recommendations for powders and solutions.
  • Used as the boronate partner in Suzuki-Miyaura cross-coupling reactions.
  • High purity (≥98.0%) suitable for research synthesis.
  • Molecular formula C13H16BNO2S and molecular weight 261.15 g/mol.
  • Available in multiple pack sizes for small-scale and preparative work.
  • Stable as a dry powder with recommended refrigerated long-term storage.
  • Suitable for use as a building block in medicinal chemistry and heterocycle synthesis.
Catalog No. 50-003-00947 Supplier Medchemexpress LLC Supplier No. HYW019633100G
Benzothiazole-5-boronic acid pinacol ester is an organoboron reagent used as the boronate partner in Suzuki-Miyaura cross-coupling reactions. It is commonly used in medicinal chemistry and synthetic organic research and should be handled and stored according to manufacturer recommendations for powders and solutions.
  • Used as the boronate partner in Suzuki-Miyaura cross-coupling reactions.
  • High purity (≥98.0%) suitable for research synthesis.
  • Molecular formula C13H16BNO2S and molecular weight 261.15 g/mol.
  • Available in multiple pack sizes for small-scale and preparative work.
  • Stable as a dry powder with recommended refrigerated long-term storage.
  • Suitable for use as a building block in medicinal chemistry and heterocycle synthesis.