Learn More
Medchemexpress LLC Benzothiazole-5-boronic acid pinacol ester | 1073354-91-2 | MFCD09260439 | >98.0% | 261.15 g·mol⁻¹ | C13H16BNO2S | 25 G
SDP
Catalog No. 5000303988
Encompass_Preferred
Benzothiazole-5-boronic acid pinacol ester is a boronate reagent used as the boronate coupling partner in Suzuki cross-coupling reactions. It is intended for research use as a synthetic building block in organic chemistry and offers a defined molecular composition and high chemical purity.
  • Suitable boronate reagent for Suzuki cross-coupling reactions.
  • High purity: ≥98.0%.
  • Molecular formula C13H16BNO2S, molecular weight 261.15 g·mol⁻¹.
  • Pinacol boronate ester offers stability under common handling conditions.
  • Supplied for research and synthetic applications.
Catalog No. 50-003-03988 Supplier Medchemexpress LLC Supplier No. 25GHYW019633
Benzothiazole-5-boronic acid pinacol ester is a boronate reagent used as the boronate coupling partner in Suzuki cross-coupling reactions. It is intended for research use as a synthetic building block in organic chemistry and offers a defined molecular composition and high chemical purity.
  • Suitable boronate reagent for Suzuki cross-coupling reactions.
  • High purity: ≥98.0%.
  • Molecular formula C13H16BNO2S, molecular weight 261.15 g·mol⁻¹.
  • Pinacol boronate ester offers stability under common handling conditions.
  • Supplied for research and synthetic applications.