Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzothiazole-5-boronic acid pinacol ester | 1073354-91-2 | MFCD09260439 | ≥98.0% | 261.15 g·mol⁻¹ | C13H16BNO2S | 50 G
SDP

Catalog No. 5000301480
Encompass_Preferred

Benzothiazole-5-boronic acid pinacol ester is a boronic ester used as the boronate coupling partner in Suzuki cross-coupling reactions. Supplied for research use, it functions as a versatile building block in organic synthesis to enable formation of carbon-carbon bonds under standard cross-coupling conditions.

  • High purity (≥98.0%) suitable for research applications.
  • Molecular formula C13H16BNO2S for accurate stoichiometry.
  • Molecular weight 261.15 g·mol⁻¹ for formulation and calculations.
  • Applicable in Suzuki cross-coupling to form aryl-aryl bonds.
  • Available in multiple package sizes to support small-scale and scale-up work.

Catalog No. 50-003-01480 Supplier Medchemexpress LLC Supplier No. HYW01963350G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzothiazole-5-boronic acid pinacol ester is a boronic ester used as the boronate coupling partner in Suzuki cross-coupling reactions. Supplied for research use, it functions as a versatile building block in organic synthesis to enable formation of carbon-carbon bonds under standard cross-coupling conditions.

  • High purity (≥98.0%) suitable for research applications.
  • Molecular formula C13H16BNO2S for accurate stoichiometry.
  • Molecular weight 261.15 g·mol⁻¹ for formulation and calculations.
  • Applicable in Suzuki cross-coupling to form aryl-aryl bonds.
  • Available in multiple package sizes to support small-scale and scale-up work.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.