Learn More
Medchemexpress LLC Benzothiazole-5-boronic acid pinacol ester | 1073354-91-2 | MFCD09260439 | >98.0% | 261.15 g·mol⁻¹ | C13H16BNO2S | 500 MG
SDP
Catalog No. 5000301479
Encompass_Preferred
Benzothiazole-5-boronic acid pinacol ester is a boronate reagent used as the boron-containing coupling partner in Suzuki-Miyaura cross-coupling reactions. Supplied as a high-purity solid for research use, it supports efficient C-C bond formation in organic synthesis and medicinal chemistry applications.
  • High purity (≥98.0%) for research applications
  • Solid form allowing straightforward handling and storage
  • Molecular formula C13H16BNO2S and molecular weight 261.15 g·mol⁻¹
  • Compatible as a boronate partner in Suzuki coupling reactions
  • Offered in multiple small-scale package sizes for lab use
Catalog No. 50-003-01479 Supplier Medchemexpress LLC Supplier No. HYW019633500MG
Benzothiazole-5-boronic acid pinacol ester is a boronate reagent used as the boron-containing coupling partner in Suzuki-Miyaura cross-coupling reactions. Supplied as a high-purity solid for research use, it supports efficient C-C bond formation in organic synthesis and medicinal chemistry applications.
  • High purity (≥98.0%) for research applications
  • Solid form allowing straightforward handling and storage
  • Molecular formula C13H16BNO2S and molecular weight 261.15 g·mol⁻¹
  • Compatible as a boronate partner in Suzuki coupling reactions
  • Offered in multiple small-scale package sizes for lab use