Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzothiazole, 6-bromo- | 53218-26-1 | 214.08 | 10 G
SDP

Catalog No. 5000445386
Encompass_Preferred

6-Bromobenzo[d]thiazole is a biochemical reagent suitable for life science research. It can be utilized as a biological material or an organic compound. This product is intended for research use only.

  • Purity of 99.0%
  • Molecular weight of 214.08
  • Appears as an off-white to yellow solid
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-45386 Supplier Medchemexpress LLC Supplier No. HYW00318210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Bromobenzo[d]thiazole is a biochemical reagent suitable for life science research. It can be utilized as a biological material or an organic compound. This product is intended for research use only.

  • Purity of 99.0%
  • Molecular weight of 214.08
  • Appears as an off-white to yellow solid
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.