Learn More
Medchemexpress LLC Benzothiazole | 95-16-9 | 99.0% | 135.19 | 100 G
SDP
Catalog No. 5000453640
Encompass_Preferred
Benzothiazole is a naturally occurring heterocyclic nuclei with various biological activities, including anticancer, antimicrobial, antidiabetic, anti-inflammatory, antileishmanial, and antiviral properties. It can also function as an efficacious β-amyloid imaging agent.
  • Naturally occurring heterocyclic nuclei.
  • Possesses various biological activities.
  • Functions as an efficacious β-amyloid imaging agent.
  • Suitable for anticancer research.
  • Suitable for antimicrobial research.
  • Suitable for antidiabetic research.
  • Suitable for anti-inflammatory research.
  • Suitable for antileishmanial research.
  • Suitable for antiviral research.
Catalog No. 50-004-53640 Supplier Medchemexpress LLC Supplier No. HYW012634100G
Benzothiazole is a naturally occurring heterocyclic nuclei with various biological activities, including anticancer, antimicrobial, antidiabetic, anti-inflammatory, antileishmanial, and antiviral properties. It can also function as an efficacious β-amyloid imaging agent.
  • Naturally occurring heterocyclic nuclei.
  • Possesses various biological activities.
  • Functions as an efficacious β-amyloid imaging agent.
  • Suitable for anticancer research.
  • Suitable for antimicrobial research.
  • Suitable for antidiabetic research.
  • Suitable for anti-inflammatory research.
  • Suitable for antileishmanial research.
  • Suitable for antiviral research.