Learn More
Medchemexpress LLC Benzothiazole | 95-16-9 | 99.04% | 135.19 | 1 ML
SDP
Catalog No. 5000453877
Encompass_Preferred
Benzothiazole is a naturally occurring heterocyclic nucleus that exhibits a variety of biological activities such as anticancer, antimicrobial, antidiabetic, anti-inflammatory, antileishmanial, and antiviral properties. Additionally, it can serve as an effective β-amyloid imaging agent.
  • Natural occurring heterocyclic nuclei
  • Possesses anticancer, antimicrobial, antidiabetic, anti-inflammatory, antileishmanial, and antiviral activities
  • Can function as an efficacious β-amyloid imaging agent
Catalog No. 50-004-53877 Supplier Medchemexpress LLC Supplier No. HYW01263410MM/1ML
Benzothiazole is a naturally occurring heterocyclic nucleus that exhibits a variety of biological activities such as anticancer, antimicrobial, antidiabetic, anti-inflammatory, antileishmanial, and antiviral properties. Additionally, it can serve as an effective β-amyloid imaging agent.
  • Natural occurring heterocyclic nuclei
  • Possesses anticancer, antimicrobial, antidiabetic, anti-inflammatory, antileishmanial, and antiviral activities
  • Can function as an efficacious β-amyloid imaging agent