Learn More
Benzothiazole, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
It is an interesting carbonyl equivalent and reacts with aldehydes or ketones to generate α-hydroxy carbonyl compounds. Benzothiazole is used as a chemical intermediate in organic synthesis. It is a precursor of rubber accelerators and a component of cyanine dyes. It is also used as a flavoring substance. It has been used as an antimicrobial agent in athletic shoe insoles. It is also used as a flavor, antimicrobial agent, and component of cyanine dyes. Its derivatives are used as rubber accelerators.
Solubility
Slightly soluble in water. Soluble in alcohol, carbon disulfide
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 2°C |
| Density | 1.241 |
| Boiling Point | 230°C |
| Flash Point | 107°C (224°F) |
| Odor | Characteristic |
| Refractive Index | 1.642 |
| Quantity | 1000 g |
| UN Number | UN2810 |
| Beilstein | 109468 |
| Merck Index | 14,1107 |
| Show More |
Safety and Handling
| Hazard Category | H302+H312-H331 |
| Hazard Statement | GHS H Statement H301-H311-H332-H319 Toxic if swallowed. Toxic in contact with skin. Harmful if inhaled. Causes serious eye irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P311-P312-P330-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S. |
| EINECSNumber | 202-396-2 |
| RTECSNumber | DL0875000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.