Learn More
Medchemexpress LLC Benzothiazolium, 2-[2-[4-[ethyl(2-hydroxyethyl)amino]phenyl]diazenyl]-6-methoxy-3-methyl-, methyl sulfate (1:1) | 12270-13-2 | 93.5% | 250 MG
SDP
Catalog No. 5000414360
Encompass_Preferred
C.I. Basic blue 41 is an alkaline dye. It can be removed from aqueous solutions using tree ferns as biosorbents in intermittent adsorption systems.
  • Alkaline dye.
  • Removable from aqueous solutions.
  • Utilizes tree ferns as biosorbents.
  • For research use only.
  • Not sold to patients.
Catalog No. 50-004-14360 Supplier Medchemexpress LLC Supplier No. 250MGHYD0492
C.I. Basic blue 41 is an alkaline dye. It can be removed from aqueous solutions using tree ferns as biosorbents in intermittent adsorption systems.
  • Alkaline dye.
  • Removable from aqueous solutions.
  • Utilizes tree ferns as biosorbents.
  • For research use only.
  • Not sold to patients.