Learn More
AdipoGen Benzothiazolyl-2EtN coumarin
SDP
Catalog No. 502561715
Encompass_Preferred
Chemical. CAS 38215-36-0. Formula C20H18N2O2S. MW 350.43. Synthetic. Fluorescent dye which is widely used for staining polymer particles, but also organelles or materials used in medicine.
Catalog No. 50-256-1715 Supplier AdipoGen Supplier No. CDXB0003G001
Chemical. CAS 38215-36-0. Formula C20H18N2O2S. MW 350.43. Synthetic. Fluorescent dye which is widely used for staining polymer particles, but also organelles or materials used in medicine.