Learn More
Medchemexpress LLC Benzothioamide-d5 | 1219804-59-7 | 142.23 | C7H2D5NS | 100 MG
SDP
Catalog No. 5000300006
Encompass_Preferred
Benzothioamide-d5 is the deuterium-labeled analogue of benzothioamide used as a stable-isotope standard and tracer for analytical and mechanistic studies. Supplied as a solid, it is intended for use as an internal standard in mass spectrometry and as a reference in NMR experiments.
  • Deuterium-labeled (d5) for clear mass shift in MS analyses
  • Suitable as an internal standard for LC-MS, GC-MS, and NMR
  • Supplied as a solid for convenient handling and storage
  • Stable under recommended storage conditions for long-term use
  • Useful as a tracer in mechanistic and quantitative studies
Catalog No. 50-003-00006 Supplier Medchemexpress LLC Supplier No. HYW015801S100MG
Benzothioamide-d5 is the deuterium-labeled analogue of benzothioamide used as a stable-isotope standard and tracer for analytical and mechanistic studies. Supplied as a solid, it is intended for use as an internal standard in mass spectrometry and as a reference in NMR experiments.
  • Deuterium-labeled (d5) for clear mass shift in MS analyses
  • Suitable as an internal standard for LC-MS, GC-MS, and NMR
  • Supplied as a solid for convenient handling and storage
  • Stable under recommended storage conditions for long-term use
  • Useful as a tracer in mechanistic and quantitative studies