Learn More
Medchemexpress LLC Benzothioamide-d5 | 1219804-59-7 | >98.0% | 142.23 g/mol | C7H2D5NS | 10 MG
SDP
Catalog No. 5000300686
Encompass_Preferred
Benzothioamide-d5 is a deuterium-labeled benzothioamide supplied as a solid powder for research use. It is intended as a tracer or internal standard to improve quantitation in analytical workflows and may be used to investigate pharmacokinetic or metabolic differences introduced by deuteration.
  • Deuterium-labeled (d5) for use as tracer or internal standard.
  • Suitable for NMR, GC-MS, and LC-MS quantitative analysis.
  • Solid powder form for ease of handling and storage.
  • Available in multiple small-mass sizes for analytical applications.
  • Stable when stored at -20 °C; in solvent stability at -80 °C for short-term storage.
Catalog No. 50-003-00686 Supplier Medchemexpress LLC Supplier No. HYW015801S10MG
Benzothioamide-d5 is a deuterium-labeled benzothioamide supplied as a solid powder for research use. It is intended as a tracer or internal standard to improve quantitation in analytical workflows and may be used to investigate pharmacokinetic or metabolic differences introduced by deuteration.
  • Deuterium-labeled (d5) for use as tracer or internal standard.
  • Suitable for NMR, GC-MS, and LC-MS quantitative analysis.
  • Solid powder form for ease of handling and storage.
  • Available in multiple small-mass sizes for analytical applications.
  • Stable when stored at -20 °C; in solvent stability at -80 °C for short-term storage.