Learn More
Medchemexpress LLC Benzothioamide-d5 | 1219804-59-7 | >98.0% | 142.23 g/mol | C7H2D5NS | 50 MG
SDP
Catalog No. 5000301540
Encompass_Preferred
Benzothioamide-d5 (CAS 1219804-59-7) is the deuterium-labeled analogue of benzothioamide supplied as a solid for research applications. It is intended as a stable-isotope reagent or analytical standard for use in mass spectrometry, isotopic tracing, and synthetic chemistry. Not for human use or clinical applications.
  • Deuterium labeled for isotopic tracing and mass spectrometry.
  • High chemical purity and isotopic enrichment (≥98%).
  • Supplied as a solid for convenient storage and handling.
  • Suitable as an internal standard for quantitative analysis.
  • For research use only; not for human or clinical use.
Catalog No. 50-003-01540 Supplier Medchemexpress LLC Supplier No. HYW015801S50MG
Benzothioamide-d5 (CAS 1219804-59-7) is the deuterium-labeled analogue of benzothioamide supplied as a solid for research applications. It is intended as a stable-isotope reagent or analytical standard for use in mass spectrometry, isotopic tracing, and synthetic chemistry. Not for human use or clinical applications.
  • Deuterium labeled for isotopic tracing and mass spectrometry.
  • High chemical purity and isotopic enrichment (≥98%).
  • Supplied as a solid for convenient storage and handling.
  • Suitable as an internal standard for quantitative analysis.
  • For research use only; not for human or clinical use.