Learn More
Medchemexpress LLC Benzothioamide-d5 | 1219804-59-7 | 99.9% | 142.23 | C7H2D5NS | 25 MG
SDP
Catalog No. 5000302906
Encompass_Preferred
Benzothioamide-d5 is a deuterium-labeled analogue of benzothioamide provided as a solid for research use. It is designed to be used as a tracer and an internal standard to support quantitative measurements in spectroscopic and mass spectrometric analyses, aiding accurate identification and quantification in laboratory assays.
  • Deuterium-labeled internal standard suitable for NMR, GC-MS, and LC-MS.
  • High chemical purity (99.9%).
  • Solid form for stable storage and handling.
  • Useful as a tracer in quantitative analyses.
  • Characterization data (SMILES, molecular weight, formula) available for confirmation.
Catalog No. 50-003-02906 Supplier Medchemexpress LLC Supplier No. 25MGHYW015801S
Benzothioamide-d5 is a deuterium-labeled analogue of benzothioamide provided as a solid for research use. It is designed to be used as a tracer and an internal standard to support quantitative measurements in spectroscopic and mass spectrometric analyses, aiding accurate identification and quantification in laboratory assays.
  • Deuterium-labeled internal standard suitable for NMR, GC-MS, and LC-MS.
  • High chemical purity (99.9%).
  • Solid form for stable storage and handling.
  • Useful as a tracer in quantitative analyses.
  • Characterization data (SMILES, molecular weight, formula) available for confirmation.