Learn More
Medchemexpress LLC HY-129943 50mg Medchemexpress, Benzothiohydrazide CAS:20605-40-7 Purity:>98%
SDP
Catalog No. 502030182
Encompass_Preferred
Medchemexpress, HY-129943 50mg Benzothiohydrazide CAS:20605-40-7 Benzothiohydrazide is an analogue of anti–tubercular agent Isoniazid. Benzothiohydrazide exhibits anti–tubercular activity, with MICs of 132 μM and 264 μM for M. tuberculosis wild type (H37Rv) and clinical mutant strains (IC1 and IC2). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0182 Supplier Medchemexpress LLC Supplier No. HY12994350MG
Medchemexpress, HY-129943 50mg Benzothiohydrazide CAS:20605-40-7 Benzothiohydrazide is an analogue of anti–tubercular agent Isoniazid. Benzothiohydrazide exhibits anti–tubercular activity, with MICs of 132 μM and 264 μM for M. tuberculosis wild type (H37Rv) and clinical mutant strains (IC1 and IC2). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.