Learn More
Medchemexpress LLC (Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate | 56602-33-6 | MFCD00011948 | ≥99.0% | 442.28 | C12H22F6N6OP2 | 250g
SDP
Catalog No. 5000267829
Encompass_Preferred
BOP (hexafluorophosphate) is a biochemical reagent that can be used as a biological material or organic compound for life science related research BOP hexafluorophosphate can be used in stepwise solid phase synthesis to prepare fragments of [Ala15]-GRF(1-29)-NH2[1]
Catalog No. 50-002-67829 Supplier Medchemexpress LLC Supplier No. HYY0973250G
BOP (hexafluorophosphate) is a biochemical reagent that can be used as a biological material or organic compound for life science related research BOP hexafluorophosphate can be used in stepwise solid phase synthesis to prepare fragments of [Ala15]-GRF(1-29)-NH2[1]