Learn More
Medchemexpress LLC Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate | 56602-33-6 | MFCD00011948 | ≥99.0% | 442.28 g·mol⁻¹ | C12H22F6N6OP2 | 100g
SDP
Catalog No. 5000268154
Encompass_Preferred
BOP (hexafluorophosphate) is a biochemical reagent that can be used as a biological material or organic compound for life science related research BOP hexafluorophosphate can be used in stepwise solid phase synthesis to prepare fragments of [Ala15]-GRF(1-29)-NH2[1]
Catalog No. 50-002-68154 Supplier Medchemexpress LLC Supplier No. HYY0973100G
BOP (hexafluorophosphate) is a biochemical reagent that can be used as a biological material or organic compound for life science related research BOP hexafluorophosphate can be used in stepwise solid phase synthesis to prepare fragments of [Ala15]-GRF(1-29)-NH2[1]