Learn More
Medchemexpress LLC Benzotriazole-5-carbonylamino methyl 3-azabicyclo[3.1.0]hexane-3-carboxylate | 2162961-71-7 | >98.0% | 527.4 g/mol | C23H19F6N5O3 | 50MG
SDP
Catalog No. 5000239472
Encompass_Preferred
BI-2545 is a potent autotaxin (ATX) inhibitor developed as a chemical probe for in vitro and in vivo research. It exhibits single-digit nanomolar potency against purified ATX and lowers plasma lysophosphatidic acid (LPA) levels in animal studies, with demonstrated oral activity in rats. The compound is provided as a solid and has reported chemical identifiers and physical properties suitable for laboratory use.
Catalog No. 50-002-39472 Supplier Medchemexpress LLC Supplier No. HY12477250MG
BI-2545 is a potent autotaxin (ATX) inhibitor developed as a chemical probe for in vitro and in vivo research. It exhibits single-digit nanomolar potency against purified ATX and lowers plasma lysophosphatidic acid (LPA) levels in animal studies, with demonstrated oral activity in rats. The compound is provided as a solid and has reported chemical identifiers and physical properties suitable for laboratory use.