Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzotriazole, Spectrum™ Chemical
SDP

Catalog No. 18620215 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
12 kg
25 kg
5 kg
500 f
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-215 500 f
18-620-214 100 g
18-620-216 5 kg
18-620-217 12 kg
18-620-218 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-620-215 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B1089500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 95-14-7

Specifications

Boiling Point 204 °C (399 °F)
Quantity 500 f
Molecular Weight (g/mol) 119.13
Percent Purity 98%
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material Benzotriazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.