Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzotrichloride, 98%

Catalog No. AAA1565236
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1565236 500 g
AAA156520E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1565236 Supplier Thermo Scientific Chemicals Supplier No. A1565236
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 98-07-7 | C7H5Cl3 | 195.47 g/mol

It finds its use in other preparations of substituted benzophenones, for benzoylation of phenols under basic, phase-transfer conditions. D,D,D-Trichlorotoluene is an intermediate, used exclusively in the industrial production of other intermediates such as Benzoylchloride, Benzotrifluoride and 2,4-Dihydroxybenzophenone. α,α,α-Trichlorotoluene is also used in determination of chlorinated toluenes in effluents of wastewater treatment plants and textile industries by closed loop stripping analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its use in other preparations of substituted benzophenones, for benzoylation of phenols under basic, phase-transfer conditions. D,D,D-Trichlorotoluene is an intermediate, used exclusively in the industrial production of other intermediates such as Benzoylchloride, Benzotrifluoride and 2,4-Dihydroxybenzophenone. α,α,α-Trichlorotoluene is also used in determination of chlorinated toluenes in effluents of wastewater treatment plants and textile industries by closed loop stripping analysis.

Solubility
Solubility in water is 0.1 g/L at 20°C.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-07-7
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.47
MDL Number MFCD00000786
InChI Key XEMRAKSQROQPBR-UHFFFAOYSA-N
Synonym benzotrichloride, trichloromethyl benzene, alpha,alpha,alpha-trichlorotoluene, phenylchloroform, toluene trichloride, benzenyl trichloride, benzene, trichloromethyl, benzyl trichloride, benzylidyne chloride, phenyltrichloromethane
PubChem CID 7367
ChEBI CHEBI:82274
IUPAC Name trichloromethylbenzene
SMILES ClC(Cl)(Cl)C1=CC=CC=C1

Specifications

Melting Point -8°C to -0.5°C
Density 1.38
Boiling Point 90°C to 91°C (15 mmHg)
Flash Point 97°C (207°F)
Refractive Index 1.557
Quantity 500 g
UN Number UN2226
Beilstein 508152
Sensitivity Moisture sensitive
Merck Index 14,1109
Formula Weight 195.48
Percent Purity 98%
Chemical Name or Material Benzotrichloride
Show More Show Less
Hazard Category H302-H315-H318-H331-H335-H350
Hazard Statement GHS H Statement
H330-H350-H314-H318-H302-H335
Fatal if inhaled.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P311-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: BENZOTRICHLORIDE
EINECSNumber 202-634-5
RTECSNumber XT9275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.