Learn More
eMolecules​ 273-53-0 | Benzoxazole | Ambeed | MFCD00005765 | 119.123 | C7H5NO | 98 | c1nc2ccccc2o1 | 25g | 525136227
Catalog No. NC2050787 Shop All eMolecules Inc. Products
Encompass_Preferred
ChemScene | 3-Hydroxy-2-methoxybenzaldehyde | 250mg | 536797322 | CS-0071433 | 66495-88-3 | MFCD06656119 | 152.149 | C8H8O3
If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool
Catalog No. NC2050787 Supplier eMolecules​ Supplier No. 525136227
ChemScene | 3-Hydroxy-2-methoxybenzaldehyde | 250mg | 536797322 | CS-0071433 | 66495-88-3 | MFCD06656119 | 152.149 | C8H8O3
If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool