Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoyl-beta-alanine 98.0+%, TCI America™
SDP

Catalog No. B00995G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B00995G 5 g
B009925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B00995G Supplier TCI America Supplier No. B00995G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3440-28-6
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
MDL Number MFCD00037293
InChI Key CWXYHOHYCJXYFQ-UHFFFAOYSA-N
Synonym Betamipron
PubChem CID 71651
IUPAC Name 3-benzamidopropanoic acid
SMILES C1=CC=C(C=C1)C(=O)NCCC(=O)O

Specifications

Melting Point 132°C
Color White
Quantity 5 g
Formula Weight 193.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzoyl-beta-alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.