Learn More
eMolecules​ 98-88-4 | Benzoyl chloride | Combi-Blocks | MFCD00000653 | 140.570 | C7H5ClO | 98.000 | ClC(=O)c1ccccc1 | 25g | 290698230
Catalog No. NC2041015 Shop All eMolecules Inc. Products
Encompass_Preferred
Benzoyl chloride | Combi-Blocks | 98-88-4 | MFCD00000653 | 140.570 | C7H5ClO | 98.000 | ClC(=O)c1ccccc1 | 25g | 290698230
Catalog No. NC2041015 Supplier eMolecules​ Supplier No. 290698230
Add to Cart
Add to Cart
Benzoyl chloride | Combi-Blocks | 98-88-4 | MFCD00000653 | 140.570 | C7H5ClO | 98.000 | ClC(=O)c1ccccc1 | 25g | 290698230