Learn More
Sigma Aldrich Fine Chemicals Biosciences Benzoyl coenzyme A lithium5MG
Catalog No. 502903113
Encompass
Benzoyl coenzyme A (Benzoyl CoA) is an intermediate in the CoA-dependent epoxide pathway synthesized from benzoate in the presence of the enzyme benzoate-CoA ligase. It is further converted to 2 3-epoxybenzoyl-CoA and this step is catalyzed by benzoyl-CoA reductase (BoxA) and benzoyl-CoA oxygenase(BoxB).
Catalog No. 50-290-3113 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. B16385MG
Benzoyl coenzyme A (Benzoyl CoA) is an intermediate in the CoA-dependent epoxide pathway synthesized from benzoate in the presence of the enzyme benzoate-CoA ligase. It is further converted to 2 3-epoxybenzoyl-CoA and this step is catalyzed by benzoyl-CoA reductase (BoxA) and benzoyl-CoA oxygenase(BoxB).