Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzoyl coenzyme A trilithium | 102185-37-5 | MFCD00078942 | 94.0% | 889.44 g·mol⁻¹ | C28H37Li3N7O17P3S | 5 MG
SDP

Catalog No. 5000262656
Encompass_Preferred

Benzoyl coenzyme A trilithium is the trilithium salt of benzoyl coenzyme A supplied for research use as an intermediate in CoA-dependent metabolic pathways (CAS 102185-37-5). It is provided as a white to off-white solid with specified storage and solvent stability for biochemical studies.

  • White to off-white solid for biochemical research.
  • Molecular weight 889.44 g·mol⁻¹.
  • Purity 94.0%.
  • Store sealed at -20°C; in solution -80°C (up to 6 months) or -20°C (up to 1 month).
  • Used to study benzoate metabolism and CoA-dependent pathways in vitro.

Catalog No. 50-002-62656 Supplier Medchemexpress LLC Supplier No. HY134129A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzoyl coenzyme A trilithium is the trilithium salt of benzoyl coenzyme A supplied for research use as an intermediate in CoA-dependent metabolic pathways (CAS 102185-37-5). It is provided as a white to off-white solid with specified storage and solvent stability for biochemical studies.

  • White to off-white solid for biochemical research.
  • Molecular weight 889.44 g·mol⁻¹.
  • Purity 94.0%.
  • Store sealed at -20°C; in solution -80°C (up to 6 months) or -20°C (up to 1 month).
  • Used to study benzoate metabolism and CoA-dependent pathways in vitro.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.