Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzoyl isocyanate

Catalog No. 3081881G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-818-81G 1 g
30-818-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30-818-81G Supplier Sigma Aldrich Supplier No. 3081881G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The reaction mechanism of benzoyl isocyanate with 6-benzyl-6-azabicyclo[2.2.1]hept-2-ene was studied.Benzoyl isocyanate was used as a capping reagent for low-reactivity hydroxy groups in the solid-phase-supported oligosaccharide synthesis.

Specifications

CAS 4461-33-0
Density 1.171 g/mL (at 25°C (literature))
Boiling Point 94°C to 96°C (21 mmHg)
Molecular Formula C8H5NO2
Refractive Index n20/D 1.5514 (literature)
Linear Formula C6H5CONCO
MDL Number MFCD00002032
Quantity 1 g
Molecular Weight (g/mol) 147.13
Percent Purity 90%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.