Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Benzoyl Peroxide
SCI_ED

Catalog No. s25672 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Powder
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25672 25 g Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25672
Only null left
Add to Cart
Edge
Add to Cart

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 94-36-0
Molecular Formula C14H10O4
Molecular Weight (g/mol) 242.23
InChI Key OMPJBNCRMGITSC-UHFFFAOYSA-N
Synonym benzoyl peroxide, dibenzoyl peroxide, peroxide, dibenzoyl, benzoperoxide, benzoyl superoxide, acetoxyl, lucidol, benoxyl, panoxyl, benzol peroxide
PubChem CID 7187
ChEBI CHEBI:82405
IUPAC Name benzoyl benzenecarboperoxoate
SMILES C1=CC=C(C=C1)C(=O)OOC(=O)C2=CC=CC=C2

Specifications

Packaging Powder
Linear Formula (C6H5CO)2O2
Quantity 25 g
Formula Weight 242.23
Chemical Name or Material Benzoyl Peroxide
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.