Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoyl(phenyliodonio)(trifluoromethanesulfonyl)methanide 98.0+%, TCI America™
SDP

Catalog No. P21435G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21435G 5 g
P21431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21435G Supplier TCI America Supplier No. P21435G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1443036-49-4
Molecular Formula C15H10F3IO3S
Molecular Weight (g/mol) 454.202
InChI Key BGLWFUSLNKHJEU-UHFFFAOYSA-N
Synonym Shibata Reagent II
PubChem CID 71813730
IUPAC Name 1-phenyl-2-(phenyl-$l^{3}-iodanylidene)-2-(trifluoromethylsulfonyl)ethanone
SMILES C1=CC=C(C=C1)C(=O)C(=IC2=CC=CC=C2)S(=O)(=O)C(F)(F)F

Specifications

Melting Point 104°C
Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 454.20
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Benzoyl(phenyliodonio)(trifluoromethanesulfonyl)methanide
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.