Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene di-p-phenyl diisocyanate, 98%, flakes

Catalog No. AC414281000
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

Catalog No. AC414281000

Supplier: Thermo Scientific Chemicals 414281000

Only null left
Add to Cart

CAS: 101-68-8 | C15H10N2O2 | 250.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-68-8, 5873-54-1
Molecular Formula C15H10N2O2
Molecular Weight (g/mol) 250.26
MDL Number MFCD00036131
InChI Key UPMLOUAZCHDJJD-UHFFFAOYSA-N
Synonym 4,4'-diphenylmethane diisocyanate, diphenylmethane diisocyanate, isonate, 4,4'-diisocyanatodiphenylmethane, p,p'-diphenylmethane diisocyanate, bis 4-isocyanatophenyl methane, 4,4'-methylenebis phenyl isocyanate, methylbisphenyl isocyanate, 1,1'-methylenebis 4-isocyanatobenzene, methylene diphenyl diisocyanate
PubChem CID 7570
ChEBI CHEBI:53218
IUPAC Name 1-isocyanato-4-[(4-isocyanatophenyl)methyl]benzene
SMILES C1=CC(=CC=C1CC2=CC=C(C=C2)N=C=O)N=C=O

Specifications

Melting Point 40.0°C
Color White
Density 1.2200g/mL
Boiling Point 392.0°C
Flash Point 202°C
Infrared Spectrum Authentic
Assay Percent Range 2.5% max. (2,4′-MDI isomer) (GC)
Packaging Glass bottle
Linear Formula OCNC6H4CH2C6H4NCO
Quantity 100 g
Specific Gravity 1.22
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in acetone,benzene,nitrobenzene
Viscosity 3-6 mPa.s (42°C)
Formula Weight 250.26
Percent Purity 98%
Physical Form Flakes
Chemical Name or Material Methylene di-p-phenyl diisocyanate, 99.5%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Suspected of causing cancer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Harmf
Precautionary Statement GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective g
EINECSNumber 202-966-0
RTECSNumber NQ9350000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.