Learn More
Medchemexpress LLC Benzoylcholine chloride | 2964-09-2 | MFCD00011786 | 98.9% | 243.73 g/mol | C12H18ClNO2 | 10 G
SDP
Catalog No. 5000282594
Encompass_Preferred
Benzoylcholine chloride is a biochemical reagent used as an organic choline derivative for biochemical assays, synthetic chemistry, and analytical studies in life-science research. Supplied as a solid powder with high purity and well-defined chemical identifiers, it is intended for laboratory research applications only.
  • High purity (98.9%) suitable for sensitive biochemical assays.
  • Molecular formula C12H18ClNO2 and molecular weight 243.73 g/mol.
  • White to almost white powder form for easy handling and dissolution.
  • Available in lab-scale sizes (5 g, 10 g, 25 g) for method development.
  • Store sealed and away from moisture; short-term cold storage recommendations for solutions.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-82594 Supplier Medchemexpress LLC Supplier No. HYW12763910G
Benzoylcholine chloride is a biochemical reagent used as an organic choline derivative for biochemical assays, synthetic chemistry, and analytical studies in life-science research. Supplied as a solid powder with high purity and well-defined chemical identifiers, it is intended for laboratory research applications only.
  • High purity (98.9%) suitable for sensitive biochemical assays.
  • Molecular formula C12H18ClNO2 and molecular weight 243.73 g/mol.
  • White to almost white powder form for easy handling and dissolution.
  • Available in lab-scale sizes (5 g, 10 g, 25 g) for method development.
  • Store sealed and away from moisture; short-term cold storage recommendations for solutions.
  • Intended for research use only; not for human or clinical use.