Learn More
Medchemexpress LLC Benzoylcholine chloride | 2964-09-2 | MFCD00011786 | 98.9% | 243.73 g/mol | C12H18ClNO2 | 25 G
SDP
Catalog No. 5000282595
Encompass_Preferred
Benzoylcholine Chloride is a quaternary ammonium biochemical reagent provided for laboratory research use. It functions as a choline/acetylcholine analog in biochemical and pharmacology assays and is supplied with quality documentation to support experimental reproducibility.
  • High purity 98.9%.
  • Molecular weight 243.73 g/mol; formula C12H18ClNO2.
  • Identified by CAS number 2964-09-2.
  • Available in 5 g, 10 g, 25 g, and 50 g package sizes.
  • Includes datasheet, certificate of analysis, safety data sheet, and handling instructions.
  • Suitable for biochemical assays and pharmacology research.
Catalog No. 50-002-82595 Supplier Medchemexpress LLC Supplier No. HYW12763925G
Benzoylcholine Chloride is a quaternary ammonium biochemical reagent provided for laboratory research use. It functions as a choline/acetylcholine analog in biochemical and pharmacology assays and is supplied with quality documentation to support experimental reproducibility.
  • High purity 98.9%.
  • Molecular weight 243.73 g/mol; formula C12H18ClNO2.
  • Identified by CAS number 2964-09-2.
  • Available in 5 g, 10 g, 25 g, and 50 g package sizes.
  • Includes datasheet, certificate of analysis, safety data sheet, and handling instructions.
  • Suitable for biochemical assays and pharmacology research.