Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzoylcholine chloride | 2964-09-2 | MFCD00011786 | 98.9% | 243.73 g/mol | C12H18ClNO2 | 5 G
SDP

Catalog No. 5000282781
Encompass_Preferred

Benzoylcholine chloride is a biochemical reagent supplied as a white to off-white solid for life-science research. It is used as an organic substrate or biological material in biochemical assays and laboratory studies, and is provided in small laboratory pack sizes with defined purity and storage recommendations.

  • CAS number 2964-09-2.
  • Molecular formula C12H18ClNO2; molecular weight 243.73 g/mol.
  • Purity 98.9%.
  • Appearance white to off-white solid.
  • Storage sealed at 4°C; in solvent: -80°C for up to 6 months, -20°C for up to 1 month.
  • Available packaging 5 g, 10 g, 25 g, 50 g.
  • Typical use substrate or reagent in biochemical assays and research.

Catalog No. 50-002-82781 Supplier Medchemexpress LLC Supplier No. HYW1276395G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzoylcholine chloride is a biochemical reagent supplied as a white to off-white solid for life-science research. It is used as an organic substrate or biological material in biochemical assays and laboratory studies, and is provided in small laboratory pack sizes with defined purity and storage recommendations.

  • CAS number 2964-09-2.
  • Molecular formula C12H18ClNO2; molecular weight 243.73 g/mol.
  • Purity 98.9%.
  • Appearance white to off-white solid.
  • Storage sealed at 4°C; in solvent: -80°C for up to 6 months, -20°C for up to 1 month.
  • Available packaging 5 g, 10 g, 25 g, 50 g.
  • Typical use substrate or reagent in biochemical assays and research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.