Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoylcholine Chloride 98.0+%, TCI America™
SDP

Catalog No. B010725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B010725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B010725G Supplier TCI America Supplier No. B010725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2964-09-2
Molecular Formula C12H18ClNO2
Molecular Weight (g/mol) 243.73
MDL Number MFCD00011786
InChI Key QVFHQENRNSAHEK-UHFFFAOYSA-M
Synonym benzoylcholine chloride, 2-hydroxyethyl trimethylammonium chloride benzoate, choline, chloride, benzoate, 2-benzoyloxy-n,n,n-trimethylethanaminium chloride, 2-benzoyloxyethyltrimethylammonium chloride, 2-benzoyloxy ethyl trimethylazanium chloride, 2-benzyloxy-n,n,n-trimethylethanaminium chloride, 2-benzyloxy-n,n,n-trimethylethanaminimum chloride, ammonium, 2-hydroxyethyl trimethyl-, chloride, benzoate, 2-trimethylamino ethyl benzoate, chloride
PubChem CID 76298
IUPAC Name 2-benzoyloxyethyl(trimethyl)azanium;chloride
SMILES C[N+](C)(C)CCOC(=O)C1=CC=CC=C1.[Cl-]

Specifications

Melting Point 208°C
Color White
Quantity 25 g
Formula Weight 243.73
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzoylcholine Chloride
RTECSNumber GA0830000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.