Learn More
Medchemexpress LLC HY-N7089 100mg Medchemexpress, Benzoyleneurea CAS:86-96-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN7089100MG
Encompass_Preferred
Medchemexpress, HY-N7089 100mg Benzoyleneurea CAS:86-96-4 Benzoyleneurea possesses anti-bacterial activity. Benzoyleneurea scaffold can be used in the synthesis of novel protein geranylgeranyltransferase-I (PGGTase-I) inhibitors [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3982