Learn More
Apexbio Technology LLC BENZOYLENEUREA-200MG
SDP
Catalog No. 5000569935
Encompass_Preferred
Benzoyleneurea (CAS No 86-96-4) is a compound of interest in biomedical research primarily within pharmacological and toxicological studies Originating from synthetic chemical processes it is known to interact with specific cellular pathways related to cell division and apoptosis although the exact targets remain under active investigation Its activity is frequently observed within the nanomolar to low micromolar range indicating potent biological effects at minimal concentrations Typical applications include the use of various cell models such as cancer cell lines as well as exploratory animal studies aimed at elucidating its pharmacokinetics and dynamics Benzoyleneurea is often employed in drug screening assays to evaluate its potential as a therapeutic agent Experimental concentrations typically depend on the specific research design allowing flexibility in its application across diverse experimental settings
Catalog No. 50-005-69935 Supplier Apexbio Technology LLC Supplier No. C7115200
Benzoyleneurea (CAS No 86-96-4) is a compound of interest in biomedical research primarily within pharmacological and toxicological studies Originating from synthetic chemical processes it is known to interact with specific cellular pathways related to cell division and apoptosis although the exact targets remain under active investigation Its activity is frequently observed within the nanomolar to low micromolar range indicating potent biological effects at minimal concentrations Typical applications include the use of various cell models such as cancer cell lines as well as exploratory animal studies aimed at elucidating its pharmacokinetics and dynamics Benzoyleneurea is often employed in drug screening assays to evaluate its potential as a therapeutic agent Experimental concentrations typically depend on the specific research design allowing flexibility in its application across diverse experimental settings